Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silicon(IV) oxide, amorphous ≥99.8% S.A. 300-350 m²/g, fumed S.A. 300-350 m²/g

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Silicium (IV) oxide amorphous Silicium dioxide amorphous Silicon dioxide amorphous

42741-A1 042741-18 42741-18 42741-A1 42741-30 42741-30
AA42741-18EA 27.39 USD InStock
CAAA42741-18 AA42741-18 CAAA42741-30 AA42741-30 CAAA42741-A1 AA42741-A1
Silicon(IV) oxide, amorphous ≥99.8% S.A. 300-350 m²/g, fumed S.A. 300-350 m²/g
Silica gel amorphous
Adhesives, coatings, cosmetics, defomers, inks, lubricants, sealants, thermal insulation


Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230 °C
Melting Pt: 1710 °C
Density: 2.2
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty