Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thymol blue sodium salt ACS

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bis[5-methyl-2-(1-methylethyl)phenolate] S,S-dioxide Thymol blue water soluble

42785-04 42785-09 042785-09
AA42785-09EA 51.69 USD InStock
AA42785-09 AA42785-04 CAAA42785-09
Thymol blue sodium salt ACS
Thymol blue sodium salt
Water soluble pH indicator


Formula: C₂₇H₂₉NaO₅S
MW: 488.58 g/mol
Melting Pt: ∼ 284 °C (dec.)
Density: 1.19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151093
CAS Number: 62625-21-2
EINECS: 263-650-6
Order Now
Specification Test Results
Clarity of solution Pass test
Visual transition interval (acid range) pH 1.2 (red) to pH 2.8 (yellow)
Visual transition interval (alkaline range) pH 8.0 (yellow) to pH 9.2 (blue)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty