Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lead ≥99.999% (metals basis), ingot

Supplier: Thermo Scientific Chemicals
Danger

043008-22 043008-36 43008-22 43008-36
AA43008-36EA 1070.99 USD InStock
AA43008-36 CAAA43008-36 AA43008-22 CAAA43008-22
Lead ≥99.999% (metals basis), ingot
Lead


Formula: Pb
MW: 207.2 g/mol
Boiling Pt: 1740 °C (1013 hPa)
Melting Pt: 327 °C
Density: 11.3437 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty