Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(2-ethoxyethyl)ether 99+% for HPLC

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Diethylene glycol diethyl ether 2-Ethoxyethyl ether

43464-AK 43464-AK
AA43464-AKEA 188.99 USD InStock
AA43464-AK CAAA43464-AK
Bis(2-ethoxyethyl)ether 99+% for HPLC
Bis(2-ethoxyethyl) ether


Formula: C₈H₁₈O₃
MW: 162.23 g/mol
Boiling Pt: 188…190 °C
Melting Pt: –44 °C
Density: 0.909
Flash Pt: 71 °C (160 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009254
CAS Number: 112-36-7
EINECS: 203-963-7
Merck Index: 13,03145
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty