Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrapropyltin

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Tetra-n-propyltin Tetrapropylstannane

71137-09 071137-09
AA71137-09EA 233.99 USD InStock
CAAA71137-09 AA71137-09
Tetrapropyltin
Tetrapropyltin


Formula: C₁₂H₂₈Sn
MW: 291.06 g/mol
Boiling Pt: 222 °C
Density: 1,11
Storage Temperature: Ambient
MDL Number: MFCD00015196
CAS Number: 2176-98-9
EINECS: 218-536-0
UN: 2788
ADR: 6.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty