Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol-[D₁] (99.5% D) (Isotopic) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Carbinol-D1 Methanol-[²H₁]

089571-14 89571-14
AA89571-14EA 109.65 USD InStock
CAAA89571-14 AA89571-14
Methanol-[D₁] (99.5% D) (Isotopic) for NMR spectroscopy
Methanol-[D1]


Formula: CH₃DO
MW: 33.03 g/mol
Boiling Pt: 64.7 °C
Melting Pt: –98 °C
Density: 0.813
Flash Pt: 11 °C (54 °F)
Storage Temperature: Ambient
MDL Number: MFCD00044213
CAS Number: 1455-13-6
EINECS: 215-933-0
UN: 1230
ADR: 3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty