Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichloroacetonitrile 98+%

Supplier: Thermo Scientific Chemicals
Danger

A10612-14 A10612-14 A10612-06 A10612-22 A10612-06 A10612-22
AAA10612-14EA 96.24 USD InStock
AAA10612-14 AAA10612-06 CAAAA10612-22 CAAAA10612-14 AAA10612-22 CAAAA10612-06
Dichloroacetonitrile 98+%
Dichloroacetonitrile


Formula: C₂HCl₂N
MW: 109.94 g/mol
Boiling Pt: 112…114 °C
Density: 1.369
Flash Pt: 35 °C(95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040886
CAS Number: 3018-12-0
EINECS: 221-159-4
UN: 2920
ADR: "8",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty