Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromobenzyl alcohol 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (3-Bromophenyl)methanol

A10929-06 A10929-14 A10929-06 A10929-14 A10929-22 A10929-22
AAA10929-06EA 41.79 USD InStock
CAAAA10929-14 CAAAA10929-06 AAA10929-06 AAAA10929-22 AAA10929-14 CAAAAA10929-22
3-Bromobenzyl alcohol 99%
3-Bromobenzyl alcohol


Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 165 °C/16 mm
Density: 1.560
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004629
CAS Number: 15852-73-0
EINECS: 239-975-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty