Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Chloropurine 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Chloro-9H-purine

A11202-03 A11202-03 A11202-06 A11202-06 A11202-14 A11202-14
AAA11202-06EA 85.69 USD InStock
AAA11202-06 CAAAA11202-06 CAAAA11202-03 CAAAA11202-14 AAA11202-03 AAA11202-14
6-Chloropurine 99%
6-Chloropurine


Formula: C₅H₃ClN₄
MW: 154.56 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00075825
CAS Number: 87-42-3
EINECS: 201-745-6
Merck Index: 13,02180
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty