Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylcyclohexanone 98+%

Supplier: Thermo Scientific Chemicals
Warning

A13186-22 A13186-36 A13186-36 A13186-22
AAA13186-22EA 129.67 USD InStock
CAAAA13186-22 CAAAA13186-36 AAA13186-22 AAA13186-36
4-Methylcyclohexanone 98+%
4-Methylcyclohexanone


Formula: C₇H₁₂O
MW: 112.17 g/mol
Boiling Pt: 169…171 °C
Melting Pt: –41 °C
Density: 0.914
Flash Pt: 48 °C(118 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001643
CAS Number: 589-92-4
EINECS: 209-665-3
UN: 2297
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty