Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylhydrazinium chloride 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Phenylhydrazine hydrochloride

A14645-22 A14645-22
AAA14645-22EA 129.47 USD InStock
CAAAA14645-22 AAA14645-22
Phenylhydrazinium chloride 99%
Phenylhydrazinium chloride


Formula: C₆H₉ClN₂
MW: 144.6 g/mol
Melting Pt: ca 252 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012924
CAS Number: 59-88-1
EINECS: 200-444-7
UN: 2811
ADR: "6.1",III
Merck Index: 12,07447
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/26/2297426.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty