Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Arginine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (R)-(-)-Arginine H-D-Arg-OH

A16137-03 A16137-06 A16137-06 A16137-03
AAA16137-06EA 267.99 USD InStock
AAA16137-06 AAA16137-03 CAAAA16137-06 CAAAA16137-03
D-(-)-Arginine 98%
D(-)-Arginine


Formula: C₆H₁₄N₄O₂
MW: 174.2 g/mol
Melting Pt: ca 235 °C
Storage Temperature: Ambient
MDL Number: MFCD00063116
CAS Number: 157-06-2
EINECS: 205-866-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty