Would you like to log in again?
KLEENGUARD® A20 Breathable Particle Protection Hoods
Supplier: Kimberly-Clark
KleenGuard®
36890 36890
10815-978CS 273.9 USD InStock
CA10815-978 10815-978
KLEENGUARD® A20 Breathable Particle Protection Hoods
Disposable Garments
Made from a patented Microforce™ Barrier SMS Fabric that offers barrier protection against dry particulates and light liquid sprays
The lightweight and breathable properties of the fabric make these hoods comfortable to wear, while the strength and abrasion-resistant properties extend the use of this product. NFPA 99 Compliant Antistatic Material.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty