Would you like to log in again?
Kimtech™ G3 Nitrile Gloves, Blue, Kimberly-Clark
Supplier: Kimberly-Clark
Kimtech™ G3
56878 56877 56879
32934-024CS 1226.35 USD InStock
32934-024 CA32934-026 CA32934-028
Kimtech™ G3 Nitrile Gloves, Blue, Kimberly-Clark
Gloves
Nitrile ambidextrous gloves are natural rubber latex-free and have a bisque finish which makes them easy to don.
  • Beaded cuff provides strength for donning and 12" length is comfortable
AQL 1.5 for pinholes and static dissipative in use.
Certifications: Double-bagged with case liner and packaged in a Class 100 Cleanroom. Recommended for ISO Class 4 or higher cleanroom environments.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty