Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzenesulfonic acid, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

M-Clarity™ quality level = MQ200

8.00468.1000 8.00468.1000
EM8.00468.1000EA 387 USD InStock
CA8.00468.1000 EM8.00468.1000
Benzenesulfonic acid, Sigma-Aldrich®
Benzenesulphonic acid


Formula: C₆H₆O₃S
MW: 158.18 g/mol
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 50 °C
Density: 1.32 g/cm³ (47 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011689
CAS Number: 98-11-3
EINECS: 202-638-7
UN: 2585
ADR: 8,III
Merck Index: 13,01070
Order Now
Specification Test Results
Assay (acidimetric) ≥ 97.0 %
Identity (IR) passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.
Sulfuric acid ≤ 0.7 %
Water (K. F.) ≤ 2.3 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty