Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dibutylamine, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger
Synonyms: Di-n-butylamineN-butyl-1-propanamine

M-Clarity™ quality level = MQ200

8.03222.1000 8.03222.1000
EM8.03222.1000EA 127 USD InStock
EM8.03222.1000 CA8.03222.1000
Dibutylamine, Sigma-Aldrich®
Dibutylamine


Formula: C₈H₁₉N
MW: 129.25 g/mol
Boiling Pt: 159 °C (1013 hPa)
Melting Pt: –62 °C
Density: 0.759 g/cm³ (20 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00009429
CAS Number: 111-92-2
EINECS: 203-921-8
UN: 2248
ADR: 8 (3),II
Merck Index: 13,03058
https://us-legacy.vwr.com/stibo/web/std.lang.all/03/15/2300315.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty