Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Gluconic acid 50% (w/w) in water

Supplier: Thermo Scientific Chemicals
Synonyms: Gluconic acid(2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanoic acid

AC11989-0010 AC11989-0100 AC11989-0025
103516-414EA 262 USD InStock
103516-414 200057-672 200057-670
D-Gluconic acid 50% (w/w) in water
D-Gluconic acid


Formula: C₆H₁₂O₇
MW: 196.16 g/mol
Melting Pt: < 15 °C
Density: 1.23 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004240
CAS Number: 526-95-4
EINECS: 208-401-4
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear yellow to brown-yellow Solution
Titration excess NaOH 49 to 55 %
Reducing sugars ≤3 %
Titration Argentometric ≥95.0 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty