Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Methylpyrrolidine 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: N-Methylpyrrolidine

AC12761-0025 AC12761-5000 AC12761-1000
103525-316EA 591 USD InStock
103525-316 200008-728 200008-730
1-Methylpyrrolidine 98%
1-Methylpyrrolidine


Formula: C₅H₁₁N
MW: 85.15 g/mol
Boiling Pt: 80…81 °C (760 mmHg)
Melting Pt: –90 °C
Density: 0.819 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00003173
CAS Number: 120-94-5
EINECS: 204-438-5
UN: 3286
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4230 to 1.4260 (20 °C, 589 nm)
GC ≥97.5 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty