Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

o-Toluonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: o-Tolunitrileo-Methylbenzonitrile2-Methylbenzonitrile

AC15286-0010 AC15286-0250 AC15286-2500
103516-506EA 592.52 USD InStock
103516-506 200013-832 200020-256
o-Toluonitrile 98%
o-Toluonitrile


Formula: C₈H₇N
MW: 117.15 g/mol
Boiling Pt: 205 °C (760 mmHg)
Melting Pt: –13 °C
Density: 0.99 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00001792
CAS Number: 529-19-1
EINECS: 208-451-7
Merck Index: 12,09675
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty