Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pentadecafluorocaprylic acid 96%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Perfluorooctanoic acidPentadecafluorooctoic acidPentadecafluorooctanoic acidPFOAPerfluorocaprylic acid

AC17396-1000 AC17396-0050 AC17396-0250
200032-460EA 116.31 USD InStock
200032-460 200026-354 102777-170
Pentadecafluorocaprylic acid 96%
Pentadecafluorocaprylic acid


Formula: C₈HF₁₅O₂
Melting Pt: 53…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00004174
CAS Number: 335-67-1
EINECS: 206-397-9
UN: 3261
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty