Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Propylene oxide 99.5%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (±)-Methyloxirane(±)-Propylene oxide(±)-Propene oxide

AC22016-0025 AC22016-0050 AC22016-0010
200026-606EA 63.11 USD InStock
200026-606 200032-708 103526-854
Propylene oxide 99.5%, Extra Pure
Propylene oxide


Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 34 °C (760 mmHg)
Melting Pt: –112 °C
Density: 0.83 g/cm³
Flash Pt: –37 °C
Storage Temperature: Ambient
MDL Number: MFCD00005126
CAS Number: 75-56-9
EINECS: 200-879-2
UN: 1280
ADR: 3,I
Merck Index: 12,08041
https://us-legacy.vwr.com/stibo/web/std.lang.all/00/14/2300014.jpg
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3650 to 1.3670 (20 °C, 589 nm)
Water ≤0.02 %
GC ≥99.4 %
Color scale ≤10 APHA
Aldehyde (C3H6O) ≤0.01 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty