Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methylindoline 99+%

Supplier: Thermo Scientific Chemicals
Warning

AC27588-0050 AC27588-1000
200041-180EA 105.86 USD InStock
200041-180 200028-878
2-Methylindoline 99+%
2-Methylindoline


Formula: C₉H₁₁N
MW: 133.19 g/mol
Boiling Pt: 227 °C (1013mbar)
Melting Pt: –51 °C
Density: 1.02 g/cm³
Flash Pt: 103 °C
MDL Number: MFCD00005706
CAS Number: 6872-06-6
EINECS: 229-971-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty