Would you like to log in again?
Puritan® Cotton Tipped Applicators, Puritan Medical Products
Supplier: Puritan Medical Products
Puritan®
25-806 2PC B 25-806 2PC Y 25-806 2PC P
89198-866CS 246.88 USD InStock
89198-866 89198-868 89198-870
Puritan® Cotton Tipped Applicators, Puritan Medical Products
Swabs Swabs for Sampling
These applicators feature high quality USP grade cotton that is bonded securely to a semi-flexible polystyrene handle by an aqueous based adhesive.
Absorbent tips are ideal for applying medicine, collecting cells, or cleaning wounds. Shorter swabs are suited for pediatric uses. Sterile. Available in three different handle colors (blue, pink, yellow). Made in USA.
Ordering information: Swabs are packaged two per envelope.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty