Would you like to log in again?
Insulated Foil Envelope Mailer, Therapak®
Supplier: Therapak
56424
89171-108CS 223.95 USD InStock
89171-108
Insulated Foil Envelope Mailer, Therapak®
Mailers and Shippers
Insulated mailer offers sanitary protection for samples transported in specimen bags.
  • Can hold multiple sample bags as a standalone mailer
  • Features a leak-resistant zipper seal and triple-layer construction to protect samples
Can also be used in conjunction with a gel wrap to form an insulating package for biological samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty