Would you like to log in again?
Westcott Fine Aspiration Biopsy Needle, BD Medical
Supplier: BD Medical
408263
BD8263BX 2485.03 USD InStock
BD8263
Westcott Fine Aspiration Biopsy Needle, BD Medical
Syringe Needles
The thin-wall biopsy needle includes a 2.2 mm slotted opening approximately 3 mm from the needle tip.
  • Two Piece
  • Thin Wall
  • Sterile
This creates a second cutting edge and enables aspirations of larger amounts of material without complications.
The two-piece needle is sterile.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty