Would you like to log in again?
IV Catheter Adapter, BD Medical
Supplier: BD Medical
403250
BD3250BX 276.11 USD InStock
BD3250
IV Catheter Adapter, BD Medical
Fittings and Connectors
Multipurpose catheter adapter provide a threaded, female BD Luer-Lok™ connection to tubing and can be used for general catheterization techniques
Tightening wings provide leverage. Sterile. Latex-free
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty