Would you like to log in again?
NORM-JECT® Catheter Sterile Syringe, Air-Tite Products
Supplier: Air-Tite
NORM-JECT®
AC50
53548-015CS 584.96 USD InStock
53548-015
NORM-JECT® Catheter Sterile Syringe, Air-Tite Products
Syringes Syringes, Medical
NORM-JECT® syringes feature medical-grade plastics, including a polypropylene barrel and a polyethylene plunger. Great value at reasonable costs.
  • Sterile
  • Certified DNA free
  • Latex free
  • Rubber free
  • Silicone free
  • Styrene free
  • DEHP free
These syringes are the choice for any situation needing an inert, non-reactive syringe. Because of their composition, these syringes are indicated for Nuclear Medicine, Amniocentisis, IVF, embryo-transfer, chromatography and many laboratory procedures. They are more chemically resistant than rubber tipped syringes. These inert syringes do not contain contaminates often associated with disposables.
The syringes can be used for sampling, mixing, and filtering. They are smooth drawing with a positive safety stop to prevent accidental spills. They feature accurate, easy-to-read increments. Their dose saver design minimizes waste, saving up to 4.5 ml of medication per 100 syringes used, as compared to non low deadspace Luer syringes (Actual savings may vary).
Certifications: Meets ISO Standards, CE Registered, FDA Registered.
Ordering information: Sterile and individually wrapped.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty