Would you like to log in again?
Dualife® Stringer Bead Twist Knot Wire Wheels, Weiler
Supplier: Weiler®
Dualife®
804-13138
300001-015EA 57.26 USD InStock
300001-015
Dualife® Stringer Bead Twist Knot Wire Wheels, Weiler
Abrasives
Stringer bead twist knot wire wheels.
  • Very narrow face
  • For hard-to-reach areas
  • Primarily for weld cleaning
  • Stainless steel wires 0.02"
These wire wheels remove rust, paint, oxides, scale, large burrs and heavy contamination.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty