Would you like to log in again?
Trulock™ Narrow-Face Crimped Wire Wheels, Weiler®
Supplier: Weiler®
Trulock™
804-01175
300010-123EA 58.22 USD InStock
300010-123
Trulock™ Narrow-Face Crimped Wire Wheels, Weiler®
Abrasives
Narrow-face crimped wire wheels.
  • Flexible brushing action
  • Can be mounted in multiples
  • Steel wires
Use these wire wheels for cleaning rust, scale and dirt, light de-burring, edge blending, roughening for adhesion, and finish preparation prior to plating or painting.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty