Would you like to log in again?
1-Minute® Gel Epoxy, Devcon
Supplier: Devcon
1-Minute®
230-14277
300050-784EA 63.37 USD InStock
300050-784
1-Minute® Gel Epoxy, Devcon
Adhesives
A multi-purpose gel epoxy.
  • Gel epoxy
  • Fills gaps
  • Non-sagging on vertical surfaces
Epoxy is fast-curing without fixturing.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty