Would you like to log in again?
Slic-Tite® Thread Tapes, PTFE, Markal®, ORS Nasco, INC.
Supplier: ORS Nasco
Slic-Tite®
434-44083
76210-664CS 109.03 USD InStock
76210-664
Slic-Tite® Thread Tapes, PTFE, Markal®, ORS Nasco, INC.
Tapes
Premium grade, high density tape.
  • Non-toxic
  • Provides a highly resistant seal on pipe threads for all types of service
  • Suitable for use with solvents, chemicals, oxygen, and on food lines
  • Permits easy disassembly, even after many years of service
  • Resistant to solvent and chemical
Applicable for metal, PVC, CPVC, ABS, and plastic. Base material is PTFE.
Pressure: 2000.00 psi
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty