Would you like to log in again?
Scotch® Glass Cloth Electrical Tapes 27, ORS Nasco, INC.
Supplier: ORS Nasco
Scotch®
500-15074
300009-340EA 50.91 USD InStock
300009-340
Scotch® Glass Cloth Electrical Tapes 27, ORS Nasco, INC.
Tapes
Scotch® Woven glass cloth tape is designed to operate in 600 V dry applications, requiring high mechanical strength and resistance to high temperatures.
  • Glass cloth is specially treated and prepared to provide corrosion-free protection
  • Will not shrink, rot, or burn
  • High tensile strength
  • Contains pressure-sensitive thermosetting adhesive which performs as Class B temperature (266 °F/130 °C)
  • Provides stable insulation for hot spot applications
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty