Would you like to log in again?
Temflex™ Vinyl Electrical Tape 1700, ORS Nasco, INC.
Supplier: ORS Nasco
Temflex™
500-69764
300009-347CS 28.31 USD InStock
300009-347
Temflex™ Vinyl Electrical Tape 1700, ORS Nasco, INC.
Tapes
Primary electrical insulation for all wire and cable splices rated up to 600 volts.
  • Polyvinyl chloride (PVC) tape is flame-retardent and conformable
  • Provides excellent mechanical protection with minimum bulk
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty