Would you like to log in again?
Crystal Simple Green®, ORS Nasco
Supplier: ORS Nasco
Simple Green®
676-19024 676-19128
300009-880CS 359.72 USD InStock
300009-880 300009-879
Crystal Simple Green®, ORS Nasco
Cleaning Agents Detergents and Surface Cleansers
  • For critical cleaning applications
  • Fragrance-free butyl cellosolve formula
  • Non-toxic, non-abrasive, and non-flammable
For critical cleaning applications that demand a fragrance-free solution. Butyl cellosolve formula is non-toxic, non-abrasive, and non-flammable.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty