Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Heavy Duty Cleaner/Degreaser, Crown
Supplier: Crown
205-8260
300050-514CS 254.33 USD InStock
300050-514
Heavy Duty Cleaner/Degreaser, Crown
Cleaning Agents Detergents and Surface Cleansers
Suitable for: Pulleys, Brakes, Engine, Alternators, Chains, Engines
  • Safe on most plastics and has a light citrus scent
  • Does not leave a residue
  • Wipes off without rinsing
  • Does not contain CFCs, HCFCs, or chlorinated solvents
  • Flash Point: 110.00 °F
Suitable for: Pulleys, Brakes, Engine, Alternators, Chains, Engines
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty