Would you like to log in again?
Mr. Clean® Finished Floor Cleaners, Procter & Gamble
Supplier: Procter & Gamble
Mr. Clean®
608-02621
300000-754PK 226.71 USD InStock
300000-754
Mr. Clean® Finished Floor Cleaners, Procter & Gamble
Cleaning Agents Detergents and Surface Cleansers
  • Patented formula removes dirt while protecting the finish
  • No-rinse cleaner leaves no dulling residue and minimizes yellowing
  • Low-sudsing formula is suitable for automatic floor scrubbing machines
  • Recommended dilution: manual (1:128), automatic machines (1:512)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty