Would you like to log in again?
All-4, CROWN, ORS Nasco
Supplier: ORS Nasco
All-4
205-7340
300050-506CS 151.87 USD InStock
300050-506
All-4, CROWN, ORS Nasco
Cleaning Agents Detergents and Surface Cleansers
Displaces moisture, lubricates and inhibits corrosion.
  • Silicone free
  • No gummy residue
  • Cleans away dirt and grease
  • Fills microscopic irregularities on surfaces
Flash Point: 155.00° F
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty