Would you like to log in again?
Crystal Clear Coatings, Krylon®
Supplier: Krylon®
425-K01301A00
300008-745CS 99.21 USD InStock
300008-745
Crystal Clear Coatings, Krylon®
Lubricants
  • Provides a tough, long-lasting protective coating
  • Guards against electrical failures
  • Will not affect resistance values or conduct current
  • Seals porous surfaces
  • Air dries in minutes
  • Adds strength and water resistance
  • Impedes fading of flourescents and other paints
  • Protects documents
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty