Would you like to log in again?
SMITH & WESSON® CALIBER™ Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
Smith & Wesson®
23006 23008 23010 23007
89130-958EA 20.06 USD InStock
89130-958 89130-954 89130-956 89130-952
SMITH & WESSON® CALIBER™ Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Safety Glasses
Ergonomic, rubber nose pads with built-in airflow holes will adjust to fit the shape of any nose for an ideal fit.
  • Contoured, soft-touch rubber temples improve wearer comfort
  • Base curve 7
  • Scratch-resistant polycarbonate lens
  • 99.9% UV protection
Cut-to-shape lens provides less distortion around the edges, and a hinge stop helps prevent lens damage.
Certifications: ANSI Z87.1
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty