Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Standard Series Hose Clamps, Precision Brand
Supplier: Precision Brand
605-35280 605-35425 605-35310 605-35250 605-35160 605-35130 605-35220
300082-686PK 95.21 USD InStock
300082-686 300082-694 300082-682 300082-692 300082-690 300082-678 300082-688
Standard Series Hose Clamps, Precision Brand
Clamps Tubing and Sanitary Clamps
Easily installed with screwdriver, nut driver, or socket wrench
  • Rounded edges to protect the hose
  • Efficient three-piece construction
  • No spot welds to corrode
Screws feature deep slots for quick installation.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty