Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Bantam Tool Clamping Tool, Band-It®, ORS Nasco
Supplier: ORS Nasco
080-C07569
300057-924EA 833.63 USD InStock
300057-924
Bantam Tool Clamping Tool, Band-It®, ORS Nasco
Pliers
Useful in tight areas with minimal access.
  • Three-way handle adjusts for one or two-handed tensioning
  • Zinc plated for corrosion resistance
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty