Would you like to log in again?
Diagonal Plastic Cutting Plier, Proto®, ORS Nasco
Supplier: ORS Nasco
577-212G
300024-144EA 59.24 USD InStock
300024-144
Diagonal Plastic Cutting Plier, Proto®, ORS Nasco
Pliers
Adjusting set screw for fast, precise cutting and preventing cutting edges from damaging each other.
  • Plastisol grips for added comfort
  • Coil spring reduces hand fatigue
  • Induction hardened cutting edges
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty