Would you like to log in again?
Proto® 13 Pc Ball-Hex L-Key Sets, ORS Nasco
Supplier: ORS Nasco
577-4995
300002-936PK 68.59 USD InStock
300002-936
Proto® 13 Pc Ball-Hex L-Key Sets, ORS Nasco
Wrenches
Set of 13 Ball-Hex L-Keys.
  • Rust resistant finish
  • Secure fastener fit
  • Heat-treated
Delivery information: Set includes 13 Hex keys per holder of different tip sizes 0.050, 1/16 , 5/64, 3/32, 7/64, 1/8, 9/64, 5/32, 3/16 , 7/32, 1/4, 5/16, 3/8".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty