Would you like to log in again?
Hi-Vis Retractable Utility Knife, Irwin®
Supplier: Irwin®
586-2082300
300004-552EA 16.83 USD InStock
300004-552
Hi-Vis Retractable Utility Knife, Irwin®
Knives
Hi-Visibility orange color makes for easy spotting on the job site.
  • Optimized cutting angle for reduced fatigue
  • Smooth, three-position slide exposes variable blade lengths
  • Screw permanently captured in casing to prevent lose
  • Inside storage for 5 blades
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty