Would you like to log in again?
Bleach Sprayers, Chapin™
Supplier: Chapin
139-20075
300037-472EA 82.8 USD InStock
300037-472
Bleach Sprayers, Chapin™
Spray Guns
Perfect for remediation and cleaning.
  • Funnel mouth opening for easy and safe filling/cleaning
  • Seals and gaskets designed to tolerate bleach solutions
  • Spray handle with lock-on spraying feature
  • Adjustable poly nozzle
Polyethylene body with plastic handle
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty