Would you like to log in again?
Stainless Steel Sprayers, Chapin™
Supplier: Chapin™
139-1749
300017-464EA 655.09 USD InStock
300017-464
Stainless Steel Sprayers, Chapin™
Spray Guns
Stainless steel sprayer with D-handle holds 3 gal.
  • Wide opening
  • No hose assembly required
  • Easy pump action for quick pressurizing
  • Adjustable cone nozzle
18" extension, 42" hose length
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty