Would you like to log in again?
Glass Tube Cutters, Wheeler-Rex
Supplier: Wheeler-Rex
828-79014
300004-962EA 161.99 USD InStock
300004-962
Glass Tube Cutters, Wheeler-Rex
Cutters
This glass tube cutters are small enough to carry in a pocket and almost as simple to use as a pair of pliers.
Cuts can be made as close as 1/2" from the end of any tubing, and in some cases even closer.
Delivery information: Supplied with chain.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty