Would you like to log in again?
Long Nose Plier with Cutter, Ampco Safety Tools
Supplier: ORS NASCO, INC.
065-P-326
300012-604EA 142.82 USD InStock
300012-604
Long Nose Plier with Cutter, Ampco Safety Tools
Pliers
Corrosion resistant.
  • Non-sparking
  • Non-magnetic
  • Beryllium-free
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty