Would you like to log in again?
Midget Tubing Cutters, Ridgid®, ORS Nasco
Supplier: ORS Nasco
Midget
632-32975
300003-650EA 33.25 USD InStock
300003-650
Midget Tubing Cutters, Ridgid®, ORS Nasco
Cutters
Designed for use in restricted spaces on small diameter, hard and soft copper, aluminum, brass, and plastic tubing
  • Strong, lightweight slide and wheel housing
  • Large knurled feed screw knob gives wasy control of cutting pressure for clean cuts
  • Screw knob hnadle
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty