Would you like to log in again?
Cushion-Grip Screwdriver Set, 8-Piece, Klein Tools, ORS Nasco
Supplier: ORS Nasco
409-85078
300004-274PK 192.95 USD InStock
300004-274
Cushion-Grip Screwdriver Set, 8-Piece, Klein Tools, ORS Nasco
Screwdrivers
Comfortable cushion-grip handles.
  • Precision-machined tips
  • Tip type and size markings
  • Flange anchors and hex-bolster caps
  • Strong, heat-treated, chrome-plated steel
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty